missing translation for 'onlineSavingsMsg'
Learn More

S100A7/Psoriasin Antibody (47C1068) - BSA Free, Novus Biologicals™

Product Code. 18779863 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18779863 0.025 mg 25µg
18706853 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18779863 Supplier Novus Biologicals Supplier No. NB10056559SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 47 publications

S100A7/Psoriasin Monoclonal specifically detects S100A7/Psoriasin in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, Flow (Intracellular).
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A7/Psoriasin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Flow Cytometry
Classification Monoclonal
Clone 47C1068
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Simple Western 1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature (PMID 26225121), Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy reported in scientific literature (PMID 26225121), Flow (Intracellular)
Gene Accession No. P31151
Gene Alias HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c
Gene Symbols S100A7
Host Species Mouse
Immunogen This antibody was developed against recombinant psoriasin/HID-5 protein.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6278
Test Specificity In confluent MCF10A cells, an 11 kDa band is observed.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.