missing translation for 'onlineSavingsMsg'
Learn More

S100A9 Antibody (CL11191) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30526349 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30526349 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30526349 Supplier Novus Biologicals Supplier No. NBP343896MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

S100A9 Monoclonal antibody specifically detects S100A9 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A9
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL11191
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B)
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of P06702, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 6280
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.