missing translation for 'onlineSavingsMsg'
Learn More

S1P2/EDG-5/S1PR2 Antibody [Janelia Fluor« 646], Novus Biologicals Biologicals™

Product Code. 30496449 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30496449 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30496449 Supplier Novus Biologicals Supplier No. NBP327900JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

S1P2/EDG-5/S1PR2 Polyclonal antibody specifically detects S1P2/EDG-5/S1PR2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen S1P2/EDG-5/S1PR2
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias AGR16, EDG5, EDG-5, endothelial differentiation G-protein coupled receptor 5, endothelial differentiation, sphingolipid G-protein-coupled receptor, 5, Gpcr13, H218, LPB2, S1P receptor 2, S1P receptor Edg-5, S1P receptor EDG5, S1P2, sphingosine 1-phosphate receptor 2, sphingosine 1-phosphate receptor Edg-5, sphingosine-1-phosphate receptor 2
Host Species Rabbit
Immunogen S1P2/EDG-5/S1PR2 antibody was raised against an 16 amino acid peptide near the carboxy terminus of human S1P2/EDG-5/S1PR2. The immunogen is located within amino acids 270 - 320 of S1P2/EDG-5/S1PR2.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 9294
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.