missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PDLIM7 Monoclonal antibody specifically detects PDLIM7 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PDLIM7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | S05-5A1 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Gene Alias | 1110003B01Rik, ENIGMAProtein enigma, LIM domain protein, LIM mineralization protein, LMP, LMP1, PDZ and LIM domain 7 (enigma), PDZ and LIM domain protein 7 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein of human PDLIM7 (Uniprot # Q9NR12) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?