missing translation for 'onlineSavingsMsg'
Learn More

Salicylic Acid 1.0 mg/ml in Acetonitrile, LoGiCal

Product Code. 30462577
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462577 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462577 Supplier LoGiCal Supplier No. LGCAMP0045.0011
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Salicylic Acid
CAS 69-72-7
Analyte or Component Names Salicylic Acid
Shipping Condition Room Temperature
Molecular Formula C7 H6 O3
Certifications/Compliance ISO 17034
Synonym Salicylic Acid, 2-Hydroxybenzoic Acid, Lamivudine Imp. C (EP), Sulfasalazine Imp. H (EP), Carbasalate Calcium Imp. C (EP), Mesalazine Imp. H (EP), Acetylsalicylic Acid Imp. C (EP), Hydroxyethyl Salicylate Imp. A (EP), Lamivudine related, Salicylic acid CCI14008X, Acetylsalicylic Acid Impurity C, Carbasalate Calcium Impurity C, Hydroxyethyl Salicylate Impurity A, Lamivudine Impurity C, Mesalazine Impurity H, Sulfasalazine Impurity H, Carbasalate Impurity C
InChI Formula InChI=1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)
Solution Type Single Solution
SMILES OC(=O)c1ccccc1O
IUPAC Name 2-hydroxybenzoic acid
Solvent or Matrix Acetonitrile
Industry Type Clinical Testing
Molecular Weight (g/mol) 138.12
Quantity 1 mL
Formula Weight 138.0317 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.