missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SAMD3 Polyclonal antibody specifically detects SAMD3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SAMD3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | bA73O6.2, FLJ34032, FLJ34563, MGC35163, SAM domain-containing protein 3, sterile alpha motif domain containing 3, sterile alpha motif domain-containing protein 3 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human SAMD3 (NP_001017373.2). METWSVEQVCSWLVEKNLGELVHRFQEEEVSGAALLALNDRMVQQLVKKIGHQAVLMDLIKKYKQNTQGLKSPENPKKAALVMQTEAARDYRDEESSSPARHGEQMPSFYPAENLDNGLIDQRVLKQRRNVKQILARSKALQWTKSYVLPEFPYDVKCMLAEQKCPDHSMRIRIIEFLQADMTKYLEGSLYPSTQQYNDVVNALLQAHPF |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?