missing translation for 'onlineSavingsMsg'
Learn More

SC65 Antibody - Azide and BSA Free, R&D Systems™

Product Code. 18611711 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18611711 0.1 mL 0.10mL
18678260 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18611711 Supplier Novus Biologicals Supplier No. NBP2946400.1ml

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

SC65 Polyclonal antibody specifically detects SC65 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SC65
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias leprecan-like 4, Leprecan-like protein 4, NO55, 55kDa, NOL55, Nucleolar autoantigen No55, SC65rat synaptonemal complex protein, synaptonemal complex protein SC65
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 338-437 of human P3H4 (NP_006446.1). FHRARWGLEEEDFQPREEAMLYHNQTAELRELLEFTHMYLQSDDEMELEETEPPLEPEDALSDAEFEGEGDYEEGMYADWWQEPDAKGDEAEAEPEPELA
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10609
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.