missing translation for 'onlineSavingsMsg'
Learn More

Schizandrin A, TRC

Product Code. 30421782
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30421782 10 mg
30422385 50 mg
30417416 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421782 Supplier Toronto Research Chemicals Supplier No. TRCS19994510MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61281-38-7
Molecular Formula C24 H32 O6
Molecular Weight (g/mol) 416.51
Synonym Dibenzo[a,c]cyclooctene, 5,6,7,8-tetrahydro-1,2,3,10,11,12-hexamethoxy-6,7-dimethyl-, (6R,7S,12aR)-, Dibenzo[a,c]cyclooctene, 5,6,7,8-tetrahydro-1,2,3,10,11,12-hexamethoxy-6,7-dimethyl-, stereoisomer, (+)-Deoxyschizandrin, (6R,7S,12aR)-5,6,7,8-Tetrahydro-1,2,3,10,11,12-hexamethoxy-6,7-dimethyldibenzo[a,c]cyclooctene, Deoxyschisandrin, Deoxyschizandrin, Schisandrin A, Schizandrin A, Wuweizisu A
SMILES COc1cc2C[C@@H](C)[C@@H](C)Cc3cc(OC)c(OC)c(OC)c3c2c(OC)c1OC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H32O6/c1-13-9-15-11-17(25-3)21(27-5)23(29-7)19(15)20-16(10-14(13)2)12-18(26-4)22(28-6)24(20)30-8/h11-14H,9-10H2,1-8H3/t13-,14+
Formula Weight 416.2199
Chemical Name or Material Schisandrin A (Deoxyschisandrin)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.