missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SCP3/SYCP3 Polyclonal specifically detects SCP3/SYCP3 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Feline, Parasite samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.
Specifications
Specifications
| Classification | Polyclonal |
| Dilution | Western Blot 0.5 ug/ml, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation reported in scientific literature (PMID 18694876), Immunohistochemistry-Paraffin 1:100 - 1:500, Immunohistochemistry-Frozen 1:200-1:500, Immunoblotting reported in scientific literature (PMID 27612028) |
| Gene Alias | anti-SCP3, MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3 |
| Purification Method | Affinity Purified |
| Target Species | Mouse, Rat, Pig, Chicken |
| Content And Storage | Store at 4C. Do not freeze. |
| Isotype | IgG |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?