missing translation for 'onlineSavingsMsg'
Learn More

SCUBE3 Antibody - BSA Free, Novus Biologicals™

Product Code. 18196448 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.03mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18196448 0.1 mg 0.1mg
18674066 0.025 mg 0.03mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18196448 Supplier Novus Biologicals Supplier No. NBP241122

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SCUBE3 Polyclonal specifically detects SCUBE3 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCUBE3
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Alias CEGF3, CUB and EGF containing protein 3, CUB domain and EGF-like repeat containing 3, DKFZp686B09105, DKFZp686B1223, DKFZp686D20108, FLJ34743, signal peptide, CUB and EGF-like domain containing protein 3, signal peptide, CUB and EGF-like domain-containing protein 3, signal peptide, CUB domain, EGF-like 3
Gene Symbols SCUBE3
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid peptide near the center of human SCUBE3. The immunogen is located within amino acids 440 - 490 of SCUBE3. Amino Acid Squence: AAPARAGHNGNSTNS
Molecular Weight of Antigen 109 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222663
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.