missing translation for 'onlineSavingsMsg'
Learn More

SDHC Antibody [PerCP], Novus Biologicals Biologicals™

Product Code. 30491904 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30491904 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30491904 Supplier Novus Biologicals Supplier No. NBP337905PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SDHC Polyclonal antibody specifically detects SDHC in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDHC
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias Integral membrane protein CII-3, mitochondrial, PGL3, QPs1, QPs-1, SDH3, succinate dehydrogenase complex, subunit C, integral membrane protein, 15kD, succinate dehydrogenase complex, subunit C, integral membrane protein, 15kDa, succinate-ubiquinone oxidoreducatase cytochrome B large subunit
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SDHC (NP_002992.1).,, Sequence:, MAALLLRHVGRHCLRAHFSPQLCIRNAVPLGTTAKEEMERFWNKNIGSNRPLSPHITIYSWSLPMAMSICHRGTGIALSAGVSLFGMSALLLPGNFESYL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6391
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.