missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SEC23A Polyclonal specifically detects SEC23A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Gene Symbols | SEC23A |
| Molecular Weight of Antigen | 86 kDa |
| Purification Method | Affinity Purified |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?