missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selegiline EP Impurity A (as Hydrochloride), Mikromol
Description
Selegiline EP Impurity A (as Hydrochloride), Mikromol
Specifications
Specifications
| Chemical Name or Material | rac-Methamphetamine hydrochloride |
| CAS | 300-42-5 |
| Analyte or Component Names | rac-Methamphetamine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H15 N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | rac-Methamphetamine Hydrochloride, N,alpha-Dimethyl-benzeneethanamine hydrochloride, N,alpha-Dimethyl-phenethylamine hydrochloride, (+/-)-Methamphetamine hydrochloride, (+/-)-N-Methylamphetamine hydrochloride, 2-Methylamino-1-phenylpropane hydrochloride, N,alpha-Dimethylphenethylamine hydrochloride, NSC 169506, NSC 22367, Obesin, Obesine, dl-Desoxyephedrine hydrochloride, dl-Methamphetamine hydrochloride, Selegiline Hydrochloride Imp. A (EP), Selegiline Imp. A (EP), Selegiline Hydrochloride Impurity A as Hydrochloride, Selegiline Impurity A as Hydrochloride |
| InChI Formula | InChI=1S/C10H15N.ClH/c1-9(11-2)8-10-6-4-3-5-7-10;/h3-7,9,11H,8H2,1-2H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CNC(C)Cc1ccccc1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?