missing translation for 'onlineSavingsMsg'
Learn More

Selegiline EP Impurity A (as Hydrochloride), Mikromol

Product Code. 30460223
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30460223 100 mg
30460832 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30460223 Supplier Mikromol Supplier No. MM0964.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Selegiline EP Impurity A (as Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-Methamphetamine hydrochloride
CAS 300-42-5
Analyte or Component Names rac-Methamphetamine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C10 H15 N . Cl H
Certifications/Compliance ISO/IEC 17025
Synonym rac-Methamphetamine Hydrochloride, N,alpha-Dimethyl-benzeneethanamine hydrochloride, N,alpha-Dimethyl-phenethylamine hydrochloride, (+/-)-Methamphetamine hydrochloride, (+/-)-N-Methylamphetamine hydrochloride, 2-Methylamino-1-phenylpropane hydrochloride, N,alpha-Dimethylphenethylamine hydrochloride, NSC 169506, NSC 22367, Obesin, Obesine, dl-Desoxyephedrine hydrochloride, dl-Methamphetamine hydrochloride, Selegiline Hydrochloride Imp. A (EP), Selegiline Imp. A (EP), Selegiline Hydrochloride Impurity A as Hydrochloride, Selegiline Impurity A as Hydrochloride
InChI Formula InChI=1S/C10H15N.ClH/c1-9(11-2)8-10-6-4-3-5-7-10;/h3-7,9,11H,8H2,1-2H3;1H
Solution Type Neat
SMILES Cl.CNC(C)Cc1ccccc1
IUPAC Name N-methyl-1-phenylpropan-2-amine;hydrochloride
Molecular Weight (g/mol) 185.69
Quantity 100 mg
Formula Weight 185.0971 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.