missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SEPHS2 Polyclonal specifically detects SEPHS2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence/Immunohistochemistry/Immunohistochemistry (Paraffin).
Specifications
Specifications
| Applications | Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Formulation | PBS |
| Gene Alias | EC 2.7.9.3, Selenophosphate synthase 2, selenophosphate synthetase 2, SPS2water dikinase 2 |
| Regulatory Status | RUO |
| Research Discipline | Core ESC Like Genes, Stem Cell Markers |
| Product Type | Antibody |
| Product Line | Novus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?