missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serine (L-Serine), LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Serine (L-Serine)
Specifications
Specifications
| Chemical Name or Material | L-Serine |
| CAS | 56-45-1 |
| Analyte or Component Names | L-Serine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C3 H7 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Serine, -2-amino-3-hydroxypropanoic acid,, (-)-Serine, -2-Amino-3-hydroxypropanoic acid, -Serine, -alpha-Amino-beta-hydroxypropionic acid, 1: PN: US20090069547 PAGE: 10 claimed protein, 225: PN: EP2071334 SEQID: 242 claimed protein, 225: PN: WO2009077864 SEQID: 242 claimed protein, 6: PN: WO2012061268 SEQID: 6 claimed protein, L-(-)-Serine, L-3-Hydroxy-2-aminopropionic acid, L-Ser, beta-Hydroxy-L-alanine |
| InChI Formula | InChI=1 S/C3H7NO3/c4-2(1-5)3(6)7/h2,5 H,1,4H2,(H,6,7)/t2-/m0/s1 |
| SMILES | N[C@@H](CO)C(=O)O |
| IUPAC Name | (2 S)-2-amino-3-hydroxypropanoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction