Learn More
Invitrogen™ Serine racemase Recombinant Rabbit Monoclonal Antibody (24GB2345)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557710
 View more versions of this product
Product Code. 30275314
  • 564.00 € / 100µL
Description
Serine racemase Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Serine racemase
Recombinant Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9GZT4
Srr
A synthesized peptide derived from human Serine racemase (aa 31-80).
100 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2345
Unconjugated
Srr
D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs
Rabbit
Affinity chromatography
RUO
63826
-20°C