missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Serine racemase Recombinant Rabbit Monoclonal Antibody (24GB2345)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

198.00 € - 564.00 €

Specifications

Antigen Serine racemase
Clone 24GB2345
Concentration 1 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275314
GREENER_CHOICE
View Documents
Invitrogen™
MA557710
100 μL
564.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275793
View Documents
Invitrogen™
MA557711
20 μL
198.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Serine racemase Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
Specifications

Specifications

Serine racemase
1 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9GZT4
63826
A synthesized peptide derived from human Serine racemase (aa 31-80).
Antibody
24GB2345
-20°C
Recombinant Monoclonal
Liquid
IgG
Human
Srr
D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs
Srr
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.