missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Serine racemase Recombinant Rabbit Monoclonal Antibody (24GB2345)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557711
This item is not returnable.
View return policy
Description
Serine racemase Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
Specifications
| Serine racemase | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9GZT4 | |
| Srr | |
| A synthesized peptide derived from human Serine racemase (aa 31-80). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2345 | |
| Unconjugated | |
| Srr | |
| D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 63826 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction