missing translation for 'onlineSavingsMsg'
Learn More

SESN3 Antibody [DyLight 680], Novus Biologicals Biologicals™

Product Code. 30503991 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30503991 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30503991 Supplier Novus Biologicals Supplier No. NBP338126FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SESN3 Polyclonal antibody specifically detects SESN3 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SESN3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias SEST3MGC29667, sestrin 3, sestrin-3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-310 of human SESN3 (NP_653266.2).,, Sequence:, MNRGGGSPSAAANYLLCTNCRKVLRKDKRIRVSQPLTRGPSAFIPEKEVVQANTVDERTNFLVEEYSTSGRLDNITQVMSLHTQYLESFLRSQFYMLRMDGPLPLPYRHYIAIMAAARHQCSYLINMHVDEFLKTGGIAEWLNGLEYVPQRLKNLNEINKLLAHRPWLITKEHIQKLVKTGENNWSLPELVHAVVLLAHYHALASFVFGSGINPERDPEISNGFRLISVNNFCVCDLANDNNIENASLSGSNFGIVDSLSELEALMERMKRLQEEREDEEASQEEMSTRFEKEKKESLFVVSGDTFHSFP
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 143686
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.