missing translation for 'onlineSavingsMsg'
Learn More

SFRS11 Rabbit, anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen SFRS11
Concentration 250 μg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Form Liquid
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18085295
View Documents
Proteintech
26751-1-AP-20UL
20 μL
145.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18089445
View Documents
Proteintech
26751-1-AP-150UL
150 μL
410.00 €
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SFRS11 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SFRS11. This antibody recognizes human, rat antigen. The SFRS11 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Specifications

SFRS11
Western Blot, Immunohistochemistry, ELISA
Rabbit
9295
IgG
Store at -20°C. Stable for one year after shipment.
250 μg/mL
Liquid
Human, Rat
p54, SFRS11, SRSF11
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.