missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTS Virus HB29 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
189.00 € - 544.00 €
Specifications
| Antigen | SFTS Virus HB29 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669797
|
Novus Biologicals
NBP2-41153-0.025mg |
0.025 mg |
189.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18124819
|
Novus Biologicals
NBP2-41153 |
0.1 mg |
544.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SFTS Virus HB29 Polyclonal specifically detects SFTS Virus HB29 in Virus samples. It is validated for Western Blot, ELISA.Specifications
| SFTS Virus HB29 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human SFTS Virus HB29 Membrane Glycoprotein. The immunogen is located within amino acids 60 - 110 of SFTS Virus HB29. Amino Acid Squence: QRGGRSQVSYYPAENSYSR | |
| Primary | |
| SFTS Virus HB29 Membrane Glycoprotein antibody is specific to the SFTS Virus HB29. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| B303_sMgp1, Membrane glycoprotein polyprotein | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title