missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SGPL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30858-100ug
This item is not returnable.
View return policy
Description
SGPL1 Polyclonal antibody specifically detects SGPL1 in Bovine,Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| SGPL1 | |
| Polyclonal | |
| Western Blot : 1:400 | |
| EC 4.1.2.27, FLJ13811, hSPL, KIAA1252, S1PL, Sphingosine-1-phosphate aldolase, sphingosine-1-phosphate lyase 1, SPLSPL 1, SP-lyase 1 | |
| Synthetic peptide derived from the SGPL1 protein. | |
| 100 μg | |
| Apoptosis | |
| 8879 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction