missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SGPL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 702.00 €
Specifications
| Antigen | SGPL1 |
|---|---|
| Dilution | Western Blot : 1:400 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066777
|
Novus Biologicals™
NBP3-30858-100ug |
100 μg |
702.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067216
|
Novus Biologicals™
NBP3-30858-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
SGPL1 Polyclonal antibody specifically detects SGPL1 in Bovine,Human,Mouse,Rat samples. It is validated for Western BlotSpecifica
| SGPL1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| Phosphate buffered saline | |
| 8879 | |
| IgG | |
| Antigen Affinity-purified |
| Western Blot : 1:400 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Human, Mouse, Rat | |
| EC 4.1.2.27, FLJ13811, hSPL, KIAA1252, S1PL, Sphingosine-1-phosphate aldolase, sphingosine-1-phosphate lyase 1, SPLSPL 1, SP-lyase 1 | |
| Synthetic peptide derived from the SGPL1 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title