missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHANK1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB300-167SS
This item is not returnable.
View return policy
Description
SHANK1 Polyclonal specifically detects SHANK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SHANK1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SHANK1 | |
| residues [SGPIYPGLFDIRSS] of the C-terminus of the SHANK1 protein. | |
| 0.025 mL | |
| Neuroscience | |
| 50944 | |
| Human, Mouse, Rat | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:250-1:750, Immunocytochemistry/Immunofluorescence 1:800, Immunohistochemistry-Paraffin 1:250-1:750 | |
| SH3 and multiple ankyrin repeat domains 1, SH3 and multiple ankyrin repeat domains protein 1, Shank1, Somatostatin receptor-interacting protein, SSTR-interacting protein, SSTRIPSPANK-1, synamon | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reacts with residues [SGPIYPGLFDIRSS] of the C-terminus of the SHANK1 protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction