missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHANK1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
226.00 € - 514.00 €
Specifications
| Antigen | SHANK1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:250-1:750, Immunocytochemistry/Immunofluorescence 1:800, Immunohistochemistry-Paraffin 1:250-1:750 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18272184
|
Novus Biologicals
NB300-167SS |
0.025 mL |
226.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18770664
|
Novus Biologicals
NB300-167 |
0.1 mL |
514.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHANK1 Polyclonal specifically detects SHANK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SHANK1 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:250-1:750, Immunocytochemistry/Immunofluorescence 1:800, Immunohistochemistry-Paraffin 1:250-1:750 | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| PBS with 0.02% Sodium Azide | |
| 50944 | |
| residues [SGPIYPGLFDIRSS] of the C-terminus of the SHANK1 protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| SH3 and multiple ankyrin repeat domains 1, SH3 and multiple ankyrin repeat domains protein 1, Shank1, Somatostatin receptor-interacting protein, SSTR-interacting protein, SSTRIPSPANK-1, synamon | |
| SHANK1 | |
| IgG | |
| Affinity Purified | |
| Reacts with residues [SGPIYPGLFDIRSS] of the C-terminus of the SHANK1 protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title