missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIAE Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
201.00 € - 470.00 €
Specifications
| Antigen | SIAE |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625770
|
Novus Biologicals
NBP2-93817-0.02ml |
0.02 mL |
201.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618501
|
Novus Biologicals
NBP2-93817-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SIAE Polyclonal antibody specifically detects SIAE in Human samples. It is validated for Western BlotSpecifications
| SIAE | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| AIS6, CSEC, CSE-C, cytosolic sialic acid 9-O-acetylesterase homolog, EC 3.1.1.53, H-Lse, LSE, MGC87009, sialate O-acetylesterase, sialic acid acetylesterase, Sialic acid-specific 9-O-acetylesterase, sialic acid-specific acetylesterase II, YSG2, Ysg2 homolog (mouse) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 270-450 of human SIAE (NP_733746.1). LKGVVWYQGESNINYNTDLYNCTFPALIEDWRETFHRGSQGQTERFFPFGLVQLSSDLSKKSSDDGFPQIRWHQTADFGYVPNPKMPNTFMAVAMDLCDRDSPFGSIHPRDKQTVAYRLHLGARALAYGEKNLTFEGPLPEKIELLAHKGLLNLTYYQQIQVQKKDNKIFEISCCSDHRCK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 54414 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title