missing translation for 'onlineSavingsMsg'
Learn More

SIAH2 Antibody (24E6H3), Novus Biologicals™

Product Code. 18758573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.125 mL
0.5 mL
Unit Size:
0.50mL
125µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18758573 0.125 mL 125µL
18757193 0.5 mL 0.50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18758573 Supplier Novus Biologicals Supplier No. NB11088113SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 17 publications

SIAH2 Monoclonal specifically detects SIAH2 in Human, Porcine, Drosophila, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIAH2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 24E6H3
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry 1:10, Immunohistochemistry 1:40-1:50, Immunocytochemistry/Immunofluorescence 1:10, Immunohistochemistry-Paraffin 1:40-1:50, Immunohistochemistry-Frozen 1:40-1:50, Flow (Intracellular)
Formulation Tissue culture supernatant with 0.1% Sodium Azide
Gene Alias E3 ubiquitin-protein ligase SIAH2, EC 6.3.2, EC 6.3.2.-, hSiah2, seven in absentia (Drosophila) homolog 2, Seven in absentia homolog 2, seven in absentia homolog 2 (Drosophila), siah-2
Gene Symbols SIAH2
Host Species Mouse
Immunogen Synthetic peptide made to an N-terminal portion of the drosophila SINA protein (within residues 1-100). [Swiss-Prot# P21461]
Purification Method Tissue culture supernatant
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Modifiers, Chromatin Research, Hypoxia, Immunology, Ubiquitin Proteasome Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6478
Test Specificity This is specific for human SIAH2 and Drosophila SINA in proliferating cells and stem cells. This does not detect Drosophila SINAH or normal, nondividing cells.
Target Species Human, Pig, Drosophila, Mouse (Negative)
Content And Storage Store at 4C. Do not freeze.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.