missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sialin/SLC17A5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00 € - 470.00 €
Specifications
| Antigen | Sialin/SLC17A5 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663242
|
Novus Biologicals
NBP2-94758-0.02ml |
0.02 mL |
188.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623952
|
Novus Biologicals
NBP2-94758-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sialin/SLC17A5 Polyclonal antibody specifically detects Sialin/SLC17A5 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| Sialin/SLC17A5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| ASTSodium/sialic acid cotransporter, FLJ22227, FLJ23268, ISSD, Membrane glycoprotein HP59, NSD, SD, sialic acid storage disease, SIALIN, SIASD, SLDSolute carrier family 17 member 5, solute carrier family 17 (anion/sugar transporter), member 5, solute carrier family 17, member 5 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SLC17A5 (NP_036566.1). MRSPVRDLARNDGEESTDRTPLLPGAPRAEAAPVCCSARYNLAILAFFGFFIVYALRVNLSVALVDMVDSNTTLEDNRTSKACPEHSAPIKVHHNQTGKK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 26503 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title