missing translation for 'onlineSavingsMsg'
Learn More

SIGLECL12 Antibody [Allophycocyanin], Novus Biologicals Biologicals™

Product Code. 30516375 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30516375 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30516375 Supplier Novus Biologicals Supplier No. NBP338557APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SIGLECL12 Polyclonal antibody specifically detects SIGLECL12 in Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLECL12
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias FLJ38600, S2V, sialic acid binding Ig-like lectin 12, sialic acid-binding Ig-like lectin 12, Siglec-12, SIGLECL1Sialic acid-binding Ig-like lectin-like 1, SIGLEC-like 1, Siglec-XII, SLGSiglec-L1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-210 of human SIGLECL12 (NP_443729.1).,, Sequence:, EQKDYLLTMQKSVTVQEGLCVSVLCSFSYPQNGWTASDPVHGYWFRAGDHVSRNIPVATNNPARAVQEETRDRFHLLGDPQNKDCTLSIRDTRESDAGTYVFCVERGNMKWNYKYDQLSVNVTASQDLLSRYRLEVPESVTVQEGLCVSVPCSVLYPHYNWTASSPVYGSWFKEGADIPWDIPVATNTPSG
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89858
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.