missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Signal sequence receptor delta Polyclonal antibody specifically detects Signal sequence receptor delta in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Signal sequence receptor delta |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:200-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | signal sequence receptor, delta (translocon-associated protein delta), SSR-delta, translocon-associated protein delta, translocon-associated protein subunit delta, TRAP-delta, TRAPDSignal sequence receptor subunit delta |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 24-144 of human SSR4 (NP_006271.1). EACLEPQITPSYYTTSDAVISTETVFIVEISLTCKNRVQNMALYADVGGKQFPVTRGQDVGRYQVSWSLDHKSAHAGTYEVRFFDEESYSLLRKAQRNNEDISIIPPLFTVSVDHRGTWNG |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?