missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIK3 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Marca: Novus Biologicals NBP2-41147-0.025mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SIK3 Polyclonal specifically detects SIK3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence.
Especificaciones
| SIK3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SIK3 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SIK3. The immunogen is located within amino acids 1190 - 1240 of SIK3. Amino Acid Squence: CHRPRALQRHHTIQNSDDA | |
| 0.025 mg | |
| Protein Kinase | |
| 23387 | |
| Human, Mouse, Rat | |
| IgY |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| EC 2.7.11, EC 2.7.11.1, KIAA0999L19, QSKFLJ12240, Salt-inducible kinase 3, Serine/threonine-protein kinase QSK, serine/threonine-protein kinase SIK3, SIK family kinase 3, SIK-3 | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least three isoforms of SIK3 are known to exist; this antibody will detect two isoforms. SIK3 antibody is predicted to not cross-react with SIK1 and SIK2. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido