missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIK3 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
178.00 € - 507.00 €
Specifications
| Antigen | SIK3 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18678545
|
Novus Biologicals
NBP2-41147-0.025mg |
0.025 mg |
178.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776893
|
Novus Biologicals
NBP2-41147 |
0.1 mg |
507.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SIK3 Polyclonal specifically detects SIK3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence.Specifications
| SIK3 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.11, EC 2.7.11.1, KIAA0999L19, QSKFLJ12240, Salt-inducible kinase 3, Serine/threonine-protein kinase QSK, serine/threonine-protein kinase SIK3, SIK family kinase 3, SIK-3 | |
| SIK3 | |
| IgY | |
| Affinity Purified |
| 1 mg/mL | |
| Polyclonal | |
| Chicken | |
| Protein Kinase | |
| PBS with 0.02% Sodium Azide | |
| 23387 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SIK3. The immunogen is located within amino acids 1190 - 1240 of SIK3. Amino Acid Squence: CHRPRALQRHHTIQNSDDA | |
| Primary | |
| At least three isoforms of SIK3 are known to exist; this antibody will detect two isoforms. SIK3 antibody is predicted to not cross-react with SIK1 and SIK2. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title