missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIK3 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41147
This item is not returnable.
View return policy
Description
SIK3 Polyclonal specifically detects SIK3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence.
Specifications
| SIK3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SIK3 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SIK3. The immunogen is located within amino acids 1190 - 1240 of SIK3. Amino Acid Squence: CHRPRALQRHHTIQNSDDA | |
| 0.1 mg | |
| Protein Kinase | |
| 23387 | |
| Human, Mouse, Rat | |
| IgY |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 20 ug/mL | |
| EC 2.7.11, EC 2.7.11.1, KIAA0999L19, QSKFLJ12240, Salt-inducible kinase 3, Serine/threonine-protein kinase QSK, serine/threonine-protein kinase SIK3, SIK family kinase 3, SIK-3 | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least three isoforms of SIK3 are known to exist; this antibody will detect two isoforms. SIK3 antibody is predicted to not cross-react with SIK1 and SIK2. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction