missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simian Immunodeficiency Virus gp120 Antibody [Allophycocyanin] - (isolate F236), Novus Biologicals™
Shop All Bio Techne ProductsDescription
Simian Immunodeficiency Virus gp120 Polyclonal antibody specifically detects Simian Immunodeficiency Virus gp120 in Virus samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Simian Immunodeficiency Virus gp120 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | APC |
| Dilution | Western Blot |
| Formulation | PBS |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Simian Immunodeficiency Virus gp120 (isolate F236) (RefSeq#: CAA32487.1; Gln24-Arg531) |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?