missing translation for 'onlineSavingsMsg'
Learn More

Simvastatin, Mikromol™

Product Code. 18032400
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18032400 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18032400 Supplier Mikromol Supplier No. MM0158.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Simvastatin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Simvastatin
CAS 79902-63-9
Analyte or Component Names Simvastatin
Shipping Condition Ice pack (-20°C)
Molecular Formula C25 H38 O5
Certifications/Compliance ISO 17025
Synonym Simvastatin, Butanoic acid, 2,2-dimethyl-, (1S,3R,7S,8S,8aR)-1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester (9CI), (+)-Simvastatin, Cholestat, Denan, Eucor, Kolestevan, L 644128-000U, Lipex, Lipinorm, Liponorm, Lipovas, Lodales, MK 733, Modutrol, Nor-Vastina, Rechol, Simcor, Simovil, Simvastatin lactone, Simvotin, Sinvacor, Sinvascor, Sivastin, Statin, Synvinolin, Valemia, Velostatin, Zocor, Zocord, Butanoic acid, 2,2-dimethyl-, 1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-(tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl)ethyl]-1-naphthalenyl ester, [1S-[1alpha,3alpha,7beta,8beta(2S*,4S*),8abeta]]-
InChI Formula InChI=1S/C25H38O5/c1-6-25(4,5)24(28)30-21-12-15(2)11-17-8-7-16(3)20(23(17)21)10-9-19-13-18(26)14-22(27)29-19/h7-8,11,15-16,18-21,23,26H,6,9-10,12-14H2,1-5H3/t15-,16-,18+,19+,20-,21-,23-/m0/s1
SMILES CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12
IUPAC Name [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbutanoate
Molecular Weight (g/mol) 418.57
Quantity 100 mg
Formula Weight 418.2719 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.