missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sin Nombre Virus Glycoprotein 2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41256
This item is not returnable.
View return policy
Description
Sin Nombre Virus Glycoprotein 2 Polyclonal specifically detects Sin Nombre Virus Glycoprotein 2 in Virus samples. It is validated for ELISA.
Specifications
| Sin Nombre Virus Glycoprotein 2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| GPC | |
| Antibody was raised against a 16 amino acid peptide near the center of the Sin Nombre virus glycoprotein. The immunogen is located within amino acids 670 - 720 of Sin Nombre Virus Glycoprotein 2. Amino Acid Squence: YSYRRKLVNPANQEET | |
| Affinity Purified | |
| RUO | |
| Virus | |
| IgG |
| ELISA | |
| 1 mg/mL | |
| ELISA 1 ug/ml | |
| Envelope polyprotein, GPC, M polyprotein, SNVsMgp1 | |
| Rabbit | |
| 126 kDa | |
| 0.1 mg | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction