missing translation for 'onlineSavingsMsg'
Learn More

Sin Nombre Virus Glycoprotein 2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

168.00 € - 507.00 €

Specifications

Antigen Sin Nombre Virus Glycoprotein 2
Concentration 1 mg/mL
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18659997
View Documents
Novus Biologicals
NBP2-41256-0.025mg
0.025 mg
168.00 €
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18194508
View Documents
Novus Biologicals
NBP2-41256
0.1 mg
507.00 €
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sin Nombre Virus Glycoprotein 2 Polyclonal specifically detects Sin Nombre Virus Glycoprotein 2 in Virus samples. It is validated for ELISA.
Specifications

Specifications

Sin Nombre Virus Glycoprotein 2
ELISA
Unconjugated
RUO
PBS with 0.02% Sodium Azide
GPC
IgG
Affinity Purified
1 mg/mL
Polyclonal
Rabbit
Virus
Envelope polyprotein, GPC, M polyprotein, SNVsMgp1
Antibody was raised against a 16 amino acid peptide near the center of the Sin Nombre virus glycoprotein. The immunogen is located within amino acids 670 - 720 of Sin Nombre Virus Glycoprotein 2. Amino Acid Squence: YSYRRKLVNPANQEET
Primary
126 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.