missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 1/SIRT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NBP2-27205SS
This item is not returnable.
View return policy
Description
Sirtuin 1/SIRT1 Polyclonal specifically detects Sirtuin 1/SIRT1 in Human, Mouse, Porcine, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Equine, Feline, Hamster, Primate, Rabbit samples. It is validated for Western Blot, Simple Western.
Specifications
| Sirtuin 1/SIRT1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| SIRT1 | |
| A portion of amino acids 400-450 from human SIRT1 was used as the immunogen for this antibody. | |
| 0.025 mg | |
| Apoptosis, Chromatin Research, DNA Repair, Epigenetics, Histone Deacetylases, Phospho Specific | |
| 23411 | |
| Human, Mouse, Pig, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Equine, Feline, Hamster, Primate, Rabbit | |
| Purified |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot 2-4ug/ml, Simple Western 1:100 | |
| EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, orangutan, chimpanzee, panda, Carolina anole, cow, new world monkey, dog, Chinese hamster, horse, chicken, mole rat, rhesus monkey, wild turkey, opossum, platypus, rabbit, pig, zebra finch, frog, cat, and white-cheeked gibbon, and 89% homologous in zebrafish, cichlid fish, and killifish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction