missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sirtuin 1/SIRT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
226.00 € - 500.00 €
Specifications
| Antigen | Sirtuin 1/SIRT1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2-4ug/ml, Simple Western 1:100 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18191114
|
Novus Biologicals
NBP2-27205SS |
0.025 mg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18736163
|
Novus Biologicals
NBP2-27205 |
0.1 mg |
500.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sirtuin 1/SIRT1 Polyclonal specifically detects Sirtuin 1/SIRT1 in Human, Mouse, Porcine, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Equine, Feline, Hamster, Primate, Rabbit samples. It is validated for Western Blot, Simple Western.Specifications
| Sirtuin 1/SIRT1 | |
| Western Blot 2-4ug/ml, Simple Western 1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Pig, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Equine, Feline, Hamster, Primate, Rabbit | |
| EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 | |
| SIRT1 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, orangutan, chimpanzee, panda, Carolina anole, cow, new world monkey, dog, Chinese hamster, horse, chicken, mole rat, rhesus monkey, wild turkey, opossum, platypus, rabbit, pig, zebra finch, frog, cat, and white-cheeked gibbon, and 89% homologous in zebrafish, cichlid fish, and killifish. |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Chromatin Research, DNA Repair, Epigenetics, Histone Deacetylases, Phospho Specific | |
| PBS with 0.05% Sodium Azide | |
| 23411 | |
| A portion of amino acids 400-450 from human SIRT1 was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title