Learn More
Sirtuin 1/SIRT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NBP2-27205
This item is not returnable.
View return policy
Description
Sirtuin 1/SIRT1 Polyclonal specifically detects Sirtuin 1/SIRT1 in Human, Mouse, Porcine, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Equine, Feline, Hamster, Primate, Rabbit samples. It is validated for Western Blot, Simple Western.
Specifications
| Polyclonal | |
| SIRT1 | |
| RUO | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 | |
| 0.1 mg | |
| Human, Mouse, Pig, Bovine, Canine, Chicken | |
| Purified |