missing translation for 'onlineSavingsMsg'
Learn More

Sisomycin sulfate, Dr. Ehrenstorfer

Product Code. 30320690
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320690 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320690 Supplier Dr. Ehrenstorfer Supplier No. DREC16970805
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sisomicin Sulfate
CAS 53179-09-2
Analyte or Component Names Sisomicin Sulfate
Shipping Condition +20°C
Molecular Formula 2 C19 H37 N5 O7 . 5 H2 O4 S
Certifications/Compliance ISO/IEC 17025
Synonym Antibiotic 66-40 sulfate, Baymicin, Extramycin, Mensiso, Sisobiotic, Sisolline, Sisomicin sulfate, Sisomin, Sisomycin sulfate, Sissomicin sulfate, D-Streptamine, O-3-deoxy-4-C-methyl-3-(methylamino)-beta-L-arabinopyranosyl-(1→6)-O-[2,6-diamino-2,3,4,6-tetradeoxy-alpha-D-glycero-hex-4-enopyranosyl-(1→4)]-2-deoxy-, sulfate (2:5), D-Streptamine, O-3-deoxy-4-C-methyl-3-(methylamino)-beta-L-arabinopyranosyl-(1→6)-O-[2,6-diamino-2,3,4,6-tetradeoxy-alpha-D-glycero-hex-4-enopyranosyl-(1→4)]-2-deoxy-, sulfate (2:5) (salt) (9CI), Gentamicin Sulfate Impurity A as Sulfate, Netilmicin Sulfate Impurity A as Sulfate, Gentamicin Impurity A as Sulfate, Netilmicin Impurity A as Sulfate
InChI Formula InChI=1S/2C19H37N5O7.5H2O4S/c2*1-19(27)7-28-18(13(26)16(19)24-2)31-15-11(23)5-10(22)14(12(15)25)30-17-9(21)4-3-8(6-20)29-17;5*1-5(2,3)4/h2*3,9-18,24-27H,4-7,20-23H2,1-2H3;5*(H2,1,2,3,4)/t2*9-,10+,11-,12+,13-,14-,15+,16-,17-,18-,19+;;;;;/m11...../s1
SMILES CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3OC(=CC[C@H]3N)CN)[C@@H]2O)OC[C@]1(C)O.CN[C@@H]4[C@@H](O)[C@@H](O[C@H]5[C@H](N)C[C@H](N)[C@@H](O[C@H]6OC(=CC[C@H]6N)CN)[C@@H]5O)OC[C@]4(C)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O
IUPAC Name (2R,3R,4R,5R)-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[[(2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid
Molecular Weight (g/mol) 1385.45
Quantity 100 mg
Formula Weight 1384.3755 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.