missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC16A4 Polyclonal antibody specifically detects SLC16A4 in Human, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SLC16A4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | MCT 5, MCT 6, MCT6, Monocarboxylate transporter 5, Monocarboxylic acid transporter 5, Monocarboxylic acid transporter 6, MOT5, SLC16A5, solute carrier family 16 (monocarboxylic acid transporters) member 5, Solute carrier family 16 member 5 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 388-487 of human SLC16A4 (NP_004687.1). TFPLLMTYTICFAIFAGGYLALILPVLVDLCRNSTVNRFLGLASFFAGMAVLSGPPIAGWLYDYTQTYNGSFYFSGICYLLSSVSFFFVPLAERWKNSLT |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?