missing translation for 'onlineSavingsMsg'
Learn More

SLC16A4 Antibody - BSA Free, Novus Biologicals™

Product Code. 18663870 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18663870 0.02 mL 0.02mL
18689810 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18663870 Supplier Novus Biologicals Supplier No. NBP2938990.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC16A4 Polyclonal antibody specifically detects SLC16A4 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC16A4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias MCT 5, MCT 6, MCT6, Monocarboxylate transporter 5, Monocarboxylic acid transporter 5, Monocarboxylic acid transporter 6, MOT5, SLC16A5, solute carrier family 16 (monocarboxylic acid transporters) member 5, Solute carrier family 16 member 5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 388-487 of human SLC16A4 (NP_004687.1). TFPLLMTYTICFAIFAGGYLALILPVLVDLCRNSTVNRFLGLASFFAGMAVLSGPPIAGWLYDYTQTYNGSFYFSGICYLLSSVSFFFVPLAERWKNSLT
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9122
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.