missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC19A3 Polyclonal antibody specifically detects SLC19A3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
Specifications
| Antigen | SLC19A3 |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, ELISA, Immunoprecipitation 1:200-1:2000 |
| Formulation | PBS, 50% Glycerol, pH (7.3) |
| Gene Alias | Solute carrier family 19 member 3, solute carrier family 19, member 3, thiamine transporter 2, ThTr2, ThTr-2 |
| Host Species | Rabbit |
| Immunogen | solute carrier family 19, member 3. (Uniprot# Q9BZV2) |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?