Learn More
Invitrogen™ SLC22A1 Recombinant Rabbit Monoclonal Antibody (24GB2510)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557256
 View more versions of this product
Product Code. 30275861
  • 146.00 € / 20µL
Description
SLC22A1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. Two transcript variants encoding two different isoforms have been found for this gene, but only the longer variant encodes a functional transporter.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC22A1
Recombinant Monoclonal
Unconjugated
SLC22A1
1-Oct; DKFZp313N1010; FLJ25043; FLJ37489; H oct1_cds; hOCT1; Lx1; mOCT1; OCT1; oct1_cds; Orct; Orct1; organic cation transporter 1; rOCT1; Slc22a1; solute carrier family 22 (organic cation transporter), member 1; solute carrier family 22 member 1; solute carrier family 22, member 1; SYK
Rabbit
Affinity chromatography
RUO
6580
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2510
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O15245
SLC22A1
A synthesized peptide derived from human SLC22A1 (aa 100-150).
20 μL