missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC22A1 Recombinant Rabbit Monoclonal Antibody (24GB2510)

Rabbit Recombinant Monoclonal Antibody

146.00 € - 500.00 €

Specifications

Antigen SLC22A1
Clone 24GB2510
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276355
View Documents
Invitrogen™
MA557255
100 μL
500.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275861
View Documents
Invitrogen™
MA557256
20 μL
146.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLC22A1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. Two transcript variants encoding two different isoforms have been found for this gene, but only the longer variant encodes a functional transporter.
Specifications

Specifications

SLC22A1
-20°C
Recombinant Monoclonal
Liquid
IgG
Human
SLC22A1
1-Oct; DKFZp313N1010; FLJ25043; FLJ37489; H oct1_cds; hOCT1; Lx1; mOCT1; OCT1; oct1_cds; Orct; Orct1; organic cation transporter 1; rOCT1; Slc22a1; solute carrier family 22 (organic cation transporter), member 1; solute carrier family 22 member 1; solute carrier family 22, member 1; SYK
SLC22A1
Primary
Affinity chromatography
24GB2510
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O15245
6580
A synthesized peptide derived from human SLC22A1 (aa 100-150).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.