missing translation for 'onlineSavingsMsg'
Learn More

SLC24A4 Antibody - BSA Free, Novus Biologicals™

Product Code. 30559153 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30559153 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30559153 Supplier Novus Biologicals Supplier No. NBP345332

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC24A4 Polyclonal antibody specifically detects SLC24A4 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC24A4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias Na(+)/K(+)/Ca(2+)-exchange protein 4, NCKX4FLJ38852, SHEP6, SLC24A2, sodium/potassium/calcium exchanger 4, solute carrier family 24 (sodium/potassium/calcium exchanger), member 4, Solute carrier family 24 member 4
Host Species Rabbit
Immunogen solute carrier family 24(sodium/potassium/calcium exchanger), member 4. (Uniprot# Q8NFF2)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123041
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.