missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A22 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94069-0.02ml
This item is not returnable.
View return policy
Description
SLC25A22 Polyclonal antibody specifically detects SLC25A22 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Gene Knock-Out
Specifications
| SLC25A22 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Knockout Validated | |
| EIEE3mitochondrial glutamate carrier 1, FLJ13044, GC1GC-1, Glutamate/H(+) symporter 1, NET44, solute carrier family 25 (mitochondrial carrier: glutamate), member 22, Solute carrier family 25 member 22 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 128-188 of human SLC25A22 (NP_078974.1). KIQLQDAGRIAAQRKILAAQGQLSAQGGAQPSVEAPAAPRPTATQLTRDLLRSRGIAGLYK | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence, Gene Knock-Out | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79751 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction