missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC28A2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93794-0.02ml
This item is not returnable.
View return policy
Description
SLC28A2 Polyclonal antibody specifically detects SLC28A2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SLC28A2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CNT2SPNT, Concentrative nucleoside transporter 2, FLJ21468, HCNT2, HsT17153, MGC138252, Na(+)/nucleoside cotransporter 2, sodium/nucleoside cotransporter 2, Sodium/purine nucleoside co-transporter, Sodium-coupled nucleoside transporter 2, solute carrier family 28 (sodium-coupled nucleoside transporter), member 2, Solute carrier family 28 member 2, SPNT1CNT 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human SLC28A2 (NP_004203.2). GAFIAFGVDASSLISASVMAAPCALASSKLAYPEVEESKFKSEEGVKLPRGKERNVLEAASNGAVDAIGLATNVAANLIAFLAVLAFINAALSWLGELVDI | |
| 0.02 mL | |
| Endocrinology, Signal Transduction | |
| 9153 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction